Learn More
Medchemexpress LLC Niclosamide (olamine) | 1420-04-8 | 98.7% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0497C100MG
Encompass_Preferred
Niclosamide olamine is an orally active antihelminthic agent used in parasitic infection research. It acts as a STAT3 inhibitor with an IC50 of 0.25 μM in HeLa cells. This compound also exhibits biological activities against cancer and suppresses DNA replication in Vero E6 cells.
  • Orally active antihelminthic agent
  • Used in parasitic infection research
  • STAT3 inhibitor
  • Exhibits biological activities against cancer
  • Inhibits DNA replication in Vero E6 cells
  • Inhibits adrenocortical carcinoma cellular proliferation
  • Inhibits STAT3-mediated luciferase reporter activity
  • Inhibits virus replication in Vero E6 cells
Catalog No. 50-004-06419