Learn More
Medchemexpress LLC Niclosamide (standard) | 50-65-7 | MFCD00057597 | 99.2% | 327.12 g·mol⁻¹ | C13H8Cl2N2O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0497R10MG
Encompass_Preferred
Niclosamide standard is an analytical reference material of the antihelminthic compound niclosamide (CAS 50-65-7) supplied as a high-purity solid for research and analytical applications. It is intended for method development, assay calibration, and qualitative or quantitative analyses using chromatography and mass spectrometry.
  • Analytical reference material for method development and calibration.
  • High purity: 99.2% as reported by the manufacturer.
  • Suitable solid form for chromatography and mass spectrometry workflows.
  • Available in small package sizes for analytical use.
  • Molecular weight 327.12 g·mol⁻¹, molecular formula C13H8Cl2N2O4.
Catalog No. 50-002-99025