Learn More
Medchemexpress LLC Nicodicosapent | 1269181-69-2 | 99.8% | 449.63 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1764050MG
Encompass_Preferred
Nicodicosapent is a fatty acid niacin conjugate that inhibits the sterol regulatory element binding protein (SREBP), a key regulator of cholesterol metabolism proteins including PCSK9, HMG-CoA reductase, ATP citrate lyase, and NPC1L1. This product is for research use only.
  • Inhibitor of sterol regulatory element binding protein (SREBP)
  • Decreases mature SREBP-2 protein production in HepG2 cells
  • Synergistically inhibits secreted PCSK9
  • Significantly inhibits ApoB secretion in a dose-dependent manner
  • Reduces PCSK9 levels, LDL particles, and plasma triglycerides in ApoE*3-Leiden mice
Catalog No. 50-003-87039