Learn More
Cayman Chemical NIcorndIl-d4 5mg
SDP
Supplier:  Cayman Chemical 336085 MG
Encompass_Preferred
An internal standard for the quantification of nicorandil by GC- or LC-MS
Catalog No. 50-295-0282