Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nicotiflorin | 17650-84-9 | MFCD03427292 | 99.9% | 594.52 | C27H30O15 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN147510MG

Encompass_Preferred

Nicotiflorin is a kaempferol-derived flavonoid glycoside extracted from Carthamus tinctorius. It exhibits antiglycation activity and has reported neuroprotective effects in preclinical studies. Supplied as a research-grade solid, it is intended for biochemical and neuropharmacology investigations.

  • High purity: 99.85% (analytical)
  • Molecular formula: C27H30O15; molecular weight: 594.52
  • Physical form: light yellow solid
  • Storage: solid at 4°C, protect from light; solutions stored at -20°C to -80°C
  • Available in small solid packs suitable for research use

Catalog No. 50-002-95710


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.