Learn More
Selleck Chemical LLC Nicotiflorin (Kaempferol-3-O-rutinoside) S3267-5mg
SDP
Supplier:  Selleck Chemical LLC S32675MG
Encompass_Preferred
Nicotiflorin (Kaempferol-3-O-rutinoside) a flavonoid extracted from Carthamus tinctorius alters the shape and structure of injured neurons decreases the number of apoptotic cells down-regulates expression of p-JAK2 p-STAT3 caspase-3 and Bax and decreases Bax immunoredactivity and increases Bcl-2 protein expression and immunoreactivity
Catalog No. 50-310-9868