Learn More
eMolecules​ NICOTINAMIDE RIBOSIDE CHLORIDE | AstaTech | 23111-00-4 | MFCD27952781 | 290.700 | C11H15ClN2O5 | 95.000 | [Cl-].NC(=O)c1ccc[n+](c1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O | 1g | 449765195
Supplier:  eMolecules​ 449765195
Encompass_Preferred
NICOTINAMIDE RIBOSIDE CHLORIDE | AstaTech | 23111-00-4 | MFCD27952781 | 290.700 | C11H15ClN2O5 | 95.000 | [Cl-].NC(=O)c1ccc[n+](c1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O | 1g | 449765195
Catalog No. NC2952134