Learn More
Selleck Chemical LLC Nicotinamide-d4 E8043-1MG
SDP
Supplier:  Selleck Chemical LLC E80431MG
Encompass_Preferred
Nicotinamide-d4 is the deuterium labeled Nicotinamide Nicotinamide is a form of vitamin B3 that plays essential roles in cell physiology through facilitating NAD redox homeostasis and providing NAD as a substrate to a class of enzymes that catalyze non-redox reactions Nicotinamide is an inhibitor of SIRT1
Catalog No. 50-000-42771