Learn More
Cayman Chemical NIcotInamIde N-oxIde 5g
Supplier: Cayman Chemical 284415 G
A metabolite of nicotinamide; formed via oxidation of nicotinamide by CYP2E1; inhibits proliferation and induces reduction of NBT in HL-60 promyelocytic leukemia cells at 30 mM; levels in urine are elevated in a mouse model of high-fat diet-induced obesity