Learn More
Selleck Chemical LLC Nicotinamide (Vitamin B3) 50mg 98-92-0 Niacinamide, Vitamin PP, Nicotinic acid amide
SDP
Supplier:  Selleck Chemical LLC S189950MG
Encompass_Preferred
Nicotinamide (Vitamin B3, Niacinamide, Vitamin PP, Nicotinic acid amide), a water-soluble vitamin, is an active component of coenzymes NAD and NADP, and also act as an inhibitor of sirtuins. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1754