Learn More
Apexbio Technology LLC Nicotinic Acid 59-67-6 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B19875.1
Encompass_Preferred
Nicotinic Acid (CAS 59-67-6) also known as niacin or vitamin B3 is a water-soluble small molecule belonging to the vitamin B complex It primarily acts by binding to and activating the G protein-coupled receptor GPR109A (also termed HCA2) expressed predominantly in adipocytes and immune cells Activation of GPR109A inhibits lipolysis reduces levels of circulating free fatty acids and modulates lipid metabolism Nicotinic acid serves as a valuable research tool in lipid-regulation studies metabolic disorders inflammation and cardiovascular biology
Catalog No. 50-000-62280