Learn More
Selleck Chemical LLC Nicotinic acid N-oxide S4995-25mg
SDP
Supplier:  Selleck Chemical LLC S499525MG
Encompass_Preferred
Nicotinic acid N-oxide (Oxiniacic acid Nicotinic acid 1-oxide) is nicotinic acid derivative that acts as a ligand and forms lead-carboxylate complexes having phosphorescent properties
Catalog No. 50-311-0932