Learn More
Hach Company Nicotinic Acid p-Toluenesulfonic Acid, 25 g
Supplier:  Hach Company 2278124
Encompass
(Nicotinic Acid PTSA) Protein-TKN standard: 4.743% N, 29.64% crude protein. For checking accuracy and digestion recovery in Total Nitrogen and TKN determinations. 25 g bottle. See 2277800 for the complete set of three standards with increasing resistance to digestion.
Catalog No. 50-199-5715