Learn More
Medchemexpress LLC Nicotinimidamide hydrochloride | 7356-60-7 | 157.60 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0022641G
Encompass_Preferred
Nicotinimidamide hydrochloride is a solid chemical compound used in laboratory settings and for the manufacture of substances. It has a molecular weight of 157.60 and a melting point of 188°C. This substance is stable under recommended storage conditions.
  • Used as a laboratory chemical
  • Stable under recommended storage conditions
  • Recommended storage at 4°C, sealed, away from moisture and light
  • Wear suitable personal protective equipment when handling
  • Avoid contact with eyes and skin
  • Ensure adequate ventilation during use
  • Keep container tightly sealed in a cool, well-ventilated area
Catalog No. 50-004-48954