Learn More
Medchemexpress LLC Nicotinuric acid | 583-08-4 | 99.9% | 180.16 | 1 ML

Supplier: Medchemexpress LLC HY11335310MM/1ML
Nicotinuric acid is an acyl glycine and a metabolite of nicotinic acid. It is classified as a human endogenous metabolite and appears as an off-white to light yellow solid.
- High purity of 99.94%
- Stable as powder for 3 years at -20°C
- Stable as powder for 2 years at 4°C
- Stable in solvent for 6 months at -80°C
- Stable in solvent for 1 month at -20°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.