Learn More
Cayman Chemical Nicotyrine 10mg
SDP
Supplier:  Cayman Chemical 1633310 MG
Encompass_Preferred
An alkaloid; reduces the number of dsDNA breaks induced by nitrosamine in a comet assay using HepaRG cells at 5 and 50 µM
Catalog No. 50-297-2643