Learn More
Bioss Niemann Pick C1 Like 1 Monoclo
SDP
Supplier:  Bioss BSM62028R
Encompass_Preferred
Niemann Pick C1 Like 1 Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-65637