Learn More
Medchemexpress LLC Niemann-Pick C2 (NPC2) protein, mouse | >95.0% | 5 UG
SDP
Supplier:  Medchemexpress LLC HYP746935UG
Encompass_Preferred
Recombinant mouse Niemann-Pick C2 (NPC2) protein expressed in HEK293 cells with a C-terminal His tag. The construct corresponds to residues E20-S149 (UniProt Q9Z0J0) and is provided as a purified preparation appropriate for biochemical assays, antibody validation, and cholesterol-binding studies. Purity is greater than 95% by reducing SDS-PAGE.
  • Expressed in HEK293 mammalian cells for native-like post-translational modifications.
  • C-terminal His tag for simplified purification and detection.
  • Amino acid range E20-S149 matching the mature protein sequence.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Suitable for biochemical assays, antibody validation, and ligand-binding studies.
  • Available in small research-scale quantities for experimental use.
Catalog No. 50-002-94495