Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Niemann-Pick C2 (NPC2) protein, recombinant (mouse) | 50 UG
SDP

Supplier:  Medchemexpress LLC HYP7469350UG

Encompass_Preferred

Niemann Pick C2 (NPC2) protein is a recombinant mouse lysosomal cholesterol-binding protein produced in HEK293 cells with a polyhistidine tag. Supplied lyophilized from PBS (pH 7.4), it is intended for biochemical and cell biology applications investigating cholesterol trafficking, lipid binding, and lysosomal function. Reconstitute and store according to manufacturer instructions to preserve activity.

  • Recombinant mouse protein expressed in HEK293 cells.
  • Polyhistidine (His) tag for affinity purification and detection.
  • Lyophilized formulation from 0.22 μm filtered PBS, pH 7.4.
  • Endotoxin level less than 1 EU/μg.
  • Molecular weight approximately 15.9-19 kDa.
  • Available in small research pack sizes, including 50 μg.
  • Reconstitute to ≥100 μg/mL per manufacturer guidance.
  • Store frozen and aliquot to avoid freeze-thaw cycles.

Catalog No. 50-002-94285


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.