Learn More
Medchemexpress LLC Nifedipine-d6 | 1188266-14-9 | 98.0% | 352.37 g/mol | C17H12D6N2O6 | 10 MG

Supplier: Medchemexpress LLC HYB0284S10MG
Nifedipine-d6 is a deuterium-labeled analogue of nifedipine supplied for analytical and research use. It is used as an internal standard or tracer in quantitative assays (NMR, GC-MS, LC-MS) and in drug metabolism and pharmacokinetic studies, providing consistent isotopic labeling for reliable measurements.
- Deuterium-labeled internal standard for quantitative assays
- Purity 98.0%
- Molecular weight 352.37 g/mol
- Molecular formula C17H12D6N2O6
- CAS number 1188266-14-9
- Storage: powder -20°C (3 years), 4°C (2 years)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.