Learn More
Cayman Chemical NIfenalolhydrochlorIde 50mg
SDP
Supplier:  Cayman Chemical 3294450 MG
Encompass_Preferred
A β-AR antagonist; inhibits isoproterenol-induced contractions in isolated rabbit right ventricular strips (pA2 = 6.05)
Catalog No. 50-294-6502