Learn More
Medchemexpress LLC Nifuroxazide | 965-52-6 | 99.9% | 1 ML

Supplier: Medchemexpress LLC HYB143610MM/1ML
Nifuroxazide is an effective inhibitor of STAT3, also exerting potent anti-tumor and anti-metastasis activity. It is an orally active nitrofuran antibiotic.
- Target is STAT3.
- Exhibits cytotoxicity against various human and NIH3T3 cell lines.
- Induces a significant dose-dependent decrease in STAT3 tyrosine phosphorylation in U266 cells.
- Causes dose-dependent loss of cell viability in U266 and INA6 cells.
- Inhibits tumor growth and weight in a dose-dependent manner.
- Inhibits proliferation of Ki-67-positive cells and induces apoptosis.
- Inhibits expression of MMP-2, MMP-9, and p-Stat3 in tumor tissues.
- Inhibits infiltration of MDSCs into the lung, suppressing distant colonization of tumor cells.
- Appearance is solid.
- Color is light yellow to yellow.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.