Learn More
Medchemexpress LLC Nifuroxazide | 965-52-6 | 99.9% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB14365G
Encompass_Preferred
Nifuroxazide is an effective inhibitor of STAT3, and also exerts potent anti-tumor and anti-metastasis activity. It is an orally active nitrofuran antibiotic.
  • Effective inhibitor of STAT3
  • Potent anti-tumor activity
  • Potent anti-metastasis activity
  • Orally active nitrofuran antibiotic
Catalog No. 50-004-12606