Learn More
Medchemexpress LLC Nifuroxazide | 965-52-6 | 99.9% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB1436500MG
Encompass_Preferred
Nifuroxazide is an orally active nitrofuran antibiotic. It is an effective inhibitor of STAT3 and also demonstrates potent anti-tumor and anti-metastasis activity. It is for research use only and not sold to patients.
  • Effective inhibitor of STAT3
  • Potent anti-tumor and anti-metastasis activity
  • Orally active nitrofuran antibiotic
  • Used in various cellular assays
  • Available in solid and solution forms
Catalog No. 50-004-13707