Learn More
Cayman Chemical NIfuroxazIde-d4 5mg
SDP
Supplier:  Cayman Chemical 333825 MG
Encompass_Preferred
An internal standard for the quantification of nifuroxazide by GC- or LC-MS
Catalog No. 50-294-6894