Learn More
Medchemexpress LLC Nifuroxazide (standard) | 965-52-6 | 10 G
SDP
Supplier:  Medchemexpress LLC HYB143610G
Encompass_Preferred
Nifuroxazide is an orally active nitrofuran antibiotic that effectively inhibits STAT3, exhibiting potent anti-tumor and anti-metastasis activities.
  • Effective inhibitor of STAT3.
  • Exhibits potent anti-tumor and anti-metastasis activity.
  • Inhibits STAT3 tyrosine phosphorylation in U266 cells.
  • Causes dose-dependent loss of cell viability in U266 or INA6 cells.
  • Inhibits tumor growth and weight in mice.
  • Inhibits proliferation of Ki-67-positive cells and induces apoptosis.
  • Inhibits expression of MMP-2, MMP-9, and p-Stat3 in A375 tumor tissues.
  • Suppresses distant colonization of tumor cells in melanoma metastasis models by inhibiting MDSCs infiltration into the lung.
Catalog No. 50-004-13705