Learn More
Selleck Chemical LLC Nigericin E4788-1G
SDP
Supplier:  Selleck Chemical LLC E47881G
Encompass_Preferred
Nigericin is an antibiotic derived from Streptomyces hygroscopicus that acts as a K /H ionophore which causes the efflux of potassium from the cell and thus influences the mitochondria membrane potential It also induces pyroptosis in macrophages by activating NLRP3 inflammasome
Catalog No. 50-000-39589