Learn More
Medchemexpress LLC Brevianamide F (cyclo(L-Pro-L-Trp)) | 38136-70-8 | MFCD29917326 | 99.0% | 283.33 g/mol | C16H17N3O2 | 10 MG

Supplier: Medchemexpress LLC HY10038510MG
Brevianamide F (Cyclo(L-Pro-L-Trp)) is a naturally occurring mycotoxin isolated from Colletotrichum gloeosporioides and provided as a characterized research reagent for biochemical and microbiological studies.
- Molecular formula C16H17N3O2 and molecular weight 283.33 g/mol.
- CAS number 38136-70-8.
- High purity reported (99.03% by HPLC).
- Available as small solid quantities (5 mg-500 mg) and 10 mM DMSO solutions.
- Supplied with data sheet, COA, SDS, and LCMS documentation.
- Reported to have antibacterial activity in the literature.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.