Learn More
Medchemexpress LLC Nigericin sodium salt | 28643-80-3 | MFCD00036825 | >=98.0% | 746.94 | C40H67NaO11 | 25 MG

Supplier: Medchemexpress LLC HY10038125MG
Nigericin sodium salt is an ionophore antibiotic derived from Streptomyces hygroscopicus that facilitates K+/H+ exchange across membranes and functions as an NLRP3 activator. It is used in research to lower intracellular pH, modulate signaling pathways, and study programmed cell death mechanisms in cancer models.
- Acts as a K+/H+ ionophore and NLRP3 activator.
- Used to decrease intracellular pH and modulate Wnt/β-catenin signaling.
- Reported to induce pyroptosis via caspase-1/GSDMD in cancer studies.
- Molecular formula C40H67NaO11; molecular weight 746.94 g/mol.
- CAS number 28643-80-3.
- High purity available (≥98% by TLC) for research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.