Learn More
Selleck Chemical LLC Nigericinsodium salt 5mg
SDP
Supplier:  Selleck Chemical LLC S66535MG
Encompass_Preferred
Nigericin (NSC 292567) sodium salt is an antibiotic derived from Streptomyces hygroscopicus that works by acting as anH+, K+, and Pb2+ ionophore.Nigericin can activateNLRP3 inflammasome to induce pro-inflammatory and immunostimulatory processes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3197