Learn More
Electron Microscopy Sciences NIGHTSEA™ Fluorescence Adapter Tru-Block Eye Shields Compact, 28-37mm
SDP
Supplier:  Electron Microscopy Sciences SFAEYEC
Encompass_Preferred
Soft, molded rubber high-sided microscope eye shields protect from light entering your eyes from the side and interfering with what you want to see under the microscope. This is very helpful in low light microscopy especailly in fluorescence microscopy. Unlike standard eye cups are available, the tall wings extend up far enough to truly shield your eyes from any level of ambient light and eliminate distractions so that you can see your subject better.Fits: 28-37mm
Catalog No. 50-192-6593