Learn More
Medchemexpress LLC Nigrosine (water soluble) | 8005-03-6 | MFCD00044681 | 100.0% | 616.49 g/mol | C22H14N6Na2O9S2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW25014525MG
Encompass_Preferred
Acid Black 2 (Nigrosine) is a water-soluble synthetic dye used as a biochemical reagent and staining agent in life-science research. Supplied as a solid powder, it is commonly used for negative staining, microscopy contrast, and color development in assays. Handle and store protected from light and under refrigerated conditions; consult the SDS for complete safety and handling guidance.
  • Water-soluble dye suitable for biological staining and microscopy.
  • Provides strong negative staining contrast for microorganisms and cells.
  • Useful for color development in biochemical assays.
  • Available in small research pack sizes for convenient laboratory use.
  • Solid powder form, readily weighed and dissolved to prepare solutions.
  • Stable when stored protected from light and refrigerated; refer to SDS for details.
  • CAS number 8005-03-6 for unambiguous chemical identification.
Catalog No. 50-002-80709