Learn More
Medchemexpress LLC Nih-12848 | 959551-10-1 | MFCD30738011 | 100.0% | 385.41 g/mol | C20H14F3N3S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10142310MG
Encompass_Preferred
NIH-12848 is a small-molecule research reagent that selectively inhibits phosphatidylinositol 5-phosphate 4-kinase γ (PI5P4Kγ) and is used in biochemical and cellular studies to probe PI5P4Kγ function and signaling pathways. For research use only; not for human or veterinary use.
  • Selective PI5P4Kγ inhibition (IC50 ≈ 1 μM).
  • No inhibition of PI5P4Kα and PI5P4Kβ at concentrations up to 100 μM.
  • High supplied purity (99.96%).
  • Solid, white to off-white powder physical form.
  • High solubility in DMSO (≥100 mg/mL).
  • Powder stable at -20°C for extended storage (years).
Catalog No. 50-002-47650