Learn More
Selleck Chemical LLC NIK SMI1 S8941-10MM/1ML
SDP
Supplier:  Selleck Chemical LLC S894110MM/1ML
Encompass_Preferred
NIK SMI1 is a highly potent and selective NF- B-inducing kinase (NIK) inhibitor with Ki of 0 23 nM for NIK-catalyzed hydrolysis of ATP to ADP
Catalog No. 50-000-24363