Learn More
Apexbio Technology LLC Nikethamide 59-26-7 500mg
SDP
Supplier:  Apexbio Technology LLC C6722500
Encompass_Preferred
Nikethamide is a small-molecule respiratory stimulant designed to modulate the respiratory center in the medulla oblongata thereby regulating central respiratory sensitivity Nikethamide exerts its biological activity primarily through modulation of neural pathways within the central nervous system potentially involving nicotinic receptors and neurotransmitter signaling cascades Based on these pharmacological properties Nikethamide holds research potential in investigating respiratory dysfunction respiratory failure and the signaling mechanisms governing respiratory neural control
Catalog No. 50-000-89223