Learn More
Medchemexpress LLC Nikkomycin Z | 59456-70-1 | 98.6% | 495.44 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY195935MG
Encompass_Preferred
Nikkomycin Z is a nucleoside peptide and an orally active antifungal agent. It inhibits chitin synthesis by acting as a competitive analogue of the chitin synthase substrate UDP-N-acetylglucosamine. Nikkomycin Z exhibits antifungal activity.
  • Inhibits the growth and biofilm formation of *Candida albicans* and *Candida parapsilosis*.
  • Shows better efficacy when used in combination with echinocandins.
  • Reduces fungal burden in a murine model of *Histoplasma capsulatum* infection.
  • Solid appearance.
  • White to off-white color.
Catalog No. 50-004-00617