Learn More
Medchemexpress LLC Nikkomycin Z | 59456-70-1 | MFCD00869648 | ≥90.0% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1959310MG
Encompass_Preferred
Nikkomycin Z is a uridine-based nucleoside-peptide antibiotic that competitively inhibits chitin synthase, producing antifungal activity and serving as a tool compound in research on fungal cell wall biosynthesis. It is supplied as a solid for mg-scale laboratory use. Chemical identifiers include CAS 59456-70-1 and molecular weight 495.4 g/mol.
  • Competitive inhibitor of chitin synthase.
  • Useful as a research tool for studying fungal cell wall biosynthesis.
  • Uridine-based nucleoside-peptide structure.
  • Provided in mg-scale solid form suitable for laboratory assays.
  • Compatible with biochemical and in vivo antifungal studies.
Catalog No. 50-002-94732