Learn More
Medchemexpress LLC Nile blue A sulfate | 3625-57-8 | MFCD00064529 | 98.0% | 732.8 g/mol | C40H40N6O6S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10190010MM 1ML
Encompass_Preferred
Nile blue A sulfate is a cationic phenoxazine dye used in histological and biochemical applications. It differentiates melanins and lipofuscins, stains lipids and fats, and can be used as a reagent in electrochemical sensor preparation. The product is commonly supplied as a ready-to-use 10 mM solution in DMSO or as a solid for laboratory research.
  • Differentiates melanin and lipofuscin in tissue samples.
  • Stains lipids and fats for histological analysis.
  • Available as a 10 mM solution in DMSO or as a solid for flexibility.
  • High purity (98.0%) suitable for research applications.
  • Useful reagent for sensor development and analytical assays.
Catalog No. 50-002-49156