Learn More
AdipoGen Nile Red
SDP
Supplier:  AdipoGen CDXN0107G005
Encompass_Preferred
Chemical. CAS 7385-67-3. Formula C20H18N2O2. MW 318.4. Synthetic. Uncharged, hydrophobic, photostable fluorescent probe that strongly fluoresces bright red in hydrophobic lipid-rich environments, but is almost nonfluorescent in water. This lipophilic stain is commonly used for the detection of intracellular lipid droplets in cells such as adipocytes by fluorescence microscopy and flow cytometry. It has been shown to detect the exposure or formation of new hydrophobic surfaces induced by ligand binding to calmodulin, oligomerization of melittin, or unfolding of ovalbumin during early thermal denaturation. This product has been employed to stain cell structures, as well as to visualize and localize colloidal drug carriers. Has been used to stain lipophilic proteins that have been separated by SDS-PAGE electrophoresis or to detect sphingolipids on thin-layer chromatograms.
Catalog No. 50-256-0411