Learn More
TARGETMOL CHEMICALS INC Nile Red 500MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T19033500MG
Encompass_Preferred
Also available in 1 g, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Nile Red (Nile Blue A oxazone) is a strongly fluorescent stain used for the detection of intracellular lipid droplets in the presence of a hydrophobic environment. Purity 95.61%
Catalog No. 50-287-8338