Learn More
Medchemexpress LLC Nile Red | 7385-67-3 | 99.7% | 159.18 UG
SDP
Supplier:  Medchemexpress LLC HYD0718Y159.18UG
Encompass_Preferred
Nile Red (Nile blue oxazone) is a lipophilic stain with environment-sensitive fluorescence. It is intensely fluorescent in a lipid-rich environment and has minimal fluorescence in aqueous media. It is an excellent vital stain for detecting intracellular lipid droplets using fluorescence microscopy and flow cytometry. Nile Red stains intracellular lipid droplets red, with a fluorescence wavelength of 559/635 nm.
  • Lipophilic stain with environment-sensitive fluorescence
  • Intensely fluorescent in a lipid-rich environment
  • Minimal fluorescence in aqueous media
  • Excellent vital stain for detecting intracellular lipid droplets
  • Stains intracellular lipid droplets red
  • Suitable for fluorescence microscopy and flow cytometry applications
  • Fluorescence wavelength of 559/635 nm
Catalog No. 50-004-10370