Learn More
Medchemexpress LLC Nilotinib | 641571-10-0 | MFCD09833716 | ≥98.0% | 529.5 g/mol | C28H22F3N7O | 500 MG
SDP
Supplier:  Medchemexpress LLC HY10159500MG
Encompass_Preferred
Nilotinib is an orally available Bcr-Abl tyrosine kinase inhibitor with antineoplastic activity used in research to study BCR-ABL-driven malignancies. It is a potent, selective small-molecule inhibitor intended for research and analytical applications.
  • Potent inhibition of Bcr-Abl tyrosine kinase.
  • Activity against several imatinib-resistant Bcr-Abl mutants.
  • Orally bioavailable small molecule suitable for in vitro and in vivo studies.
  • High chemical purity suitable for analytical and research use.
  • Defined chemical identity: C28H22F3N7O, MW 529.5 g/mol.
Catalog No. 50-202-8672