Learn More
Apexbio Technology LLC Nilotinib(AMN-107), 1g. Cas: 641571-10-0 MFCD: MFCD09833716. Bcr-Abl kinase inhibitor,selective
SDP
Supplier:  Apexbio Technology LLC A82321000
Encompass_Preferred
Nilotinib is a selective, oral inhibitor of tyrosine kinase with IC50 values of 20, 42, 31, 38, 29 and 41 nM for the autophosphorylation of native BCR-ABL (WT p210) and mutant BCR-ABL (E281K, E292K, F317L, M351T and F486S), respectively [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5084