Learn More
Selleck Chemical LLC Nilotinib hydrochloride S5205-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S520510MM/1ML
Encompass_Preferred
Nilotinib hydrochloride (AMN-107) is the hydrochloride salt form of nilotinib an orally bioavailable Bcr-Abl tyrosine kinase inhibitor with antineoplastic activity
Catalog No. 50-311-2256