Learn More
Medchemexpress LLC Nilutamide-d6 | 1189477-66-4 | 99.8% | 323.26 g/mol | C12H4D6F3N3O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY13702S5MG
Encompass_Preferred
Nilutamide-d6 is the deuterium-labeled isotopologue of nilutamide supplied as a stable isotope reference standard for analytical and research applications. It is intended for use as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS in studies of androgen receptor pharmacology. For research use only; not for human or clinical use.
  • High isotopic labeling (D6) for reliable internal standardization.
  • Suitable as tracer and internal standard for NMR, GC-MS, and LC-MS.
  • High purity (99.8%) for accurate quantitation.
  • Available in small mg pack sizes for analytical workflows.
Catalog No. 50-002-22411