Learn More
Cayman Chemical NIlvadIpIn-d4 1mg
SDP
Supplier:  Cayman Chemical 337001 MG
Encompass_Preferred
An internal standard for the quantification of nilvadipine by GC- or LC-MS
Catalog No. 50-294-8033