Learn More
Apexbio Technology LLC Nilvadipine 75530-68-6 50mg
SDP
Supplier:  Apexbio Technology LLC A845550
Encompass_Preferred
Nilvadipine (CAS 75530-68-6) is a dihydropyridine calcium channel antagonist that inhibits L-type voltage-gated calcium channels in vascular smooth muscle By reducing transmembrane calcium influx Nilvadipine induces vasodilation and lowers blood pressure making it widely studied in hypertension research In preclinical and clinical studies it has been employed to investigate vascular physiology neuroprotection and cardiovascular pharmacology supporting its use as a probe in the evaluation of calcium channel blockade and associated therapeutic strategies
Catalog No. 50-000-95832