Learn More
Selleck Chemical LLC Nimorazole
SDP
Supplier:  Selleck Chemical LLC S641125MG
Encompass_Preferred
Nimorazole is a water soluble 5-nitroimidazole compound with antibacterial and potential radiosensitizing activity
Catalog No. 50-000-04778