Learn More
Medchemexpress LLC Ningetinib Tosylate 10mM 1mL | 1394820-77-9 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY10714510MM 1ML
Encompass_Preferred
Ningetinib Tosylate is a potent orally bioavailable small molecule tyrosine kinase inhibitor (TKI) with IC50s of 6 7 1 9 and 1 0 nM for c-Met VEGFR2 and Axl respectively
Catalog No. 50-002-40877