Learn More
Alkali Scientific Ninhydrin Reagent, 10 G
Supplier: Alkali Scientific NIN002CP
Ninhydrin Reagent, 10g is commonly used in amino acid detection, particularly in chromatography, where it reacts with free amino acids to produce a purple color. The reaction is widely used to quantify and identify amino acids in biological samples, making it essential in proteomics and biochemical analysis. Ninhydrin is used extensively in clinical diagnostics, forensics, and research laboratories to analyze protein content and detect amino acids in samples such as urine, plasma, and blood. This 10g pack size is ideal for smaller-scale testing or routine laboratory use, providing a cost-effective solution for experiments that require frequent application of the reagent.